Learn More
Medchemexpress LLC Tetrahydrozoline hydrochloride (Tetryzoline hydrochloride) | 522-48-5 | 99.9% | 1 ML
SDP
Catalog No. 5000409459
Encompass_Preferred
Tetrahydrozoline hydrochloride is an alpha-adrenergic agonist and a derivative of imidazoline that causes vasoconstriction. It is widely used in research for nasal congestion and conjunctival congestion, and induces the synthesis of collagen types I and III in primary human gingival fibroblasts. This product is for research use only.
  • Causes vasoconstriction
  • Used in research for nasal congestion
  • Used in research for conjunctival congestion
  • Induces the synthesis of collagen types I and III in primary human gingival fibroblasts
  • Functions as an alpha-adrenergic agonist
Catalog No. 50-004-09459 Supplier Medchemexpress LLC Supplier No. HYB0556A10MM/1ML
Tetrahydrozoline hydrochloride is an alpha-adrenergic agonist and a derivative of imidazoline that causes vasoconstriction. It is widely used in research for nasal congestion and conjunctival congestion, and induces the synthesis of collagen types I and III in primary human gingival fibroblasts. This product is for research use only.
  • Causes vasoconstriction
  • Used in research for nasal congestion
  • Used in research for conjunctival congestion
  • Induces the synthesis of collagen types I and III in primary human gingival fibroblasts
  • Functions as an alpha-adrenergic agonist