Learn More
Medchemexpress LLC Tetraiodothyroacetic acid | 67-30-1 | >99.2% | 747.83 | 5 MG
SDP
Catalog No. 5000451303
Encompass_Preferred
Tetrac (Tetraiodothyroacetic acid) is a derivative of L-thyroxine (T4) and acts as a thyrointegrin receptor antagonist. It blocks the actions of T4 and 3,5,3'-triiodo-L-thyronine (T3) at the cell surface receptor for thyroid hormone on integrin αvβ3.
  • Exhibits anti-angiogenic activity.
  • Exhibits anti-tumor activity.
Catalog No. 50-004-51303 Supplier Medchemexpress LLC Supplier No. HYW0088595MG
Tetrac (Tetraiodothyroacetic acid) is a derivative of L-thyroxine (T4) and acts as a thyrointegrin receptor antagonist. It blocks the actions of T4 and 3,5,3'-triiodo-L-thyronine (T3) at the cell surface receptor for thyroid hormone on integrin αvβ3.
  • Exhibits anti-angiogenic activity.
  • Exhibits anti-tumor activity.