Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraisobutyl Orthotitanate (contains Isopropoxide), TCI America™
SDP

Catalog No. T097825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T097825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T097825ML Supplier TCI America Supplier No. T097825ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7425-80-1
Molecular Formula C16H36O4Ti
Molecular Weight (g/mol) 340.327
MDL Number MFCD00015045
InChI Key QUVMSYUGOKEMPX-UHFFFAOYSA-N
Synonym Isobutyl Titanate, Titanium Isobutoxide
PubChem CID 165601
IUPAC Name 2-methylpropan-1-olate;titanium(4+)
SMILES CC(C)C[O-].CC(C)C[O-].CC(C)C[O-].CC(C)C[O-].[Ti+4]

Specifications

Color Yellow
Quantity 25 mL
UN Number 1993
Formula Weight 340.33
Physical Form Liquid
Chemical Name or Material Tetraisobutyl Orthotitanate (contains Isopropoxide)
EINECSNumber (2)-0228&(2)-2150
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.