Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraisobutyl Orthotitanate, TCI America™
SDP

Catalog No. T27115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27115G Supplier TCI America Supplier No. T27115G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7425-80-1
Molecular Formula C16H36O4Ti
Molecular Weight (g/mol) 340.327
MDL Number MFCD00015045
InChI Key QUVMSYUGOKEMPX-UHFFFAOYSA-N
Synonym Isobutyl Titanate, Titanium Isobutoxide
PubChem CID 165601
IUPAC Name 2-methylpropan-1-olate;titanium(4+)
SMILES CC(C)C[O-].CC(C)C[O-].CC(C)C[O-].CC(C)C[O-].[Ti+4]

Specifications

Melting Point 40°C
Color White-Yellow
Boiling Point 141°C
Quantity 5 g
Formula Weight 340.33
Physical Form Crystalline Powder
Chemical Name or Material Tetraisobutyl Orthotitanate
EINECSNumber (2)-0228&(2)-2150
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.