Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraisobutylene (contains isomer), TCI America™
SDP

Catalog No. T013225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T013225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T013225ML Supplier TCI America Supplier No. T013225ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15220-85-6
Molecular Formula C16H32
Molecular Weight (g/mol) 224.432
MDL Number MFCD00059946
InChI Key URRHKOYTHDCSDA-UHFFFAOYSA-N
PubChem CID 56846471
IUPAC Name 2,5,8,11-tetramethyldodec-2-ene
SMILES CC(C)CCC(C)CCC(C)CC=C(C)C

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 224.43
Physical Form Liquid
Chemical Name or Material Tetraisobutylene (contains isomer)
EINECSNumber (2)-0032&(2)-0027
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.