Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(1,1,1,3,3,3-hexafluoroisopropyl) Orthosilicate, TCI America™
SDP

Catalog No. T25821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T25821G Supplier TCI America Supplier No. T25821G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26560-90-7
Molecular Formula C12H4F24O4Si
Molecular Weight (g/mol) 696.207
InChI Key BBAPGYPPYPFUOP-UHFFFAOYSA-N
Synonym Tetrakis(1,1,1,3,3,3-hexafluoroisopropoxy)silane
PubChem CID 11542124
IUPAC Name tetrakis(1,1,1,3,3,3-hexafluoropropan-2-yl) silicate
SMILES C(C(F)(F)F)(C(F)(F)F)O[Si](OC(C(F)(F)F)C(F)(F)F)(OC(C(F)(F)F)C(F)(F)F)OC(C(F)(F)F)C(F)(F)F

Specifications

Color Colorless
Boiling Point 58°C
UN Number 2924
Quantity 1 g
Formula Weight 696.21
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrakis(1,1,1,3,3,3-hexafluoroisopropyl) Orthosilicate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.