Learn More
Strem, An Ascensus Company CAS# 14221-01-3. Tetrakis(triphenylphosphine)palladium(0), 99% (99.9+%-Pd) MFCD00010012. 25G
SDP
Catalog No. 501959477
Encompass
CAS# 14221-01-3. 25G. Tetrakis(triphenylphosphine)palladium(0), 99% (99.9+%-Pd). MFCD00010012. Molecular Formula: C72H60P4Pd. Molecular Weight: 1155.57. Color/Form: bright yellow pwdr. Stability: air sensitive, store cold (-18°C) Strem#: 46-2150 www.strem.com/catalog/v3/46-2150/
Catalog No. 50-195-9477 Supplier Strem, An Ascensus Company Supplier No. 46215025G
CAS# 14221-01-3. 25G. Tetrakis(triphenylphosphine)palladium(0), 99% (99.9+%-Pd). MFCD00010012. Molecular Formula: C72H60P4Pd. Molecular Weight: 1155.57. Color/Form: bright yellow pwdr. Stability: air sensitive, store cold (-18°C) Strem#: 46-2150 www.strem.com/catalog/v3/46-2150/