Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(2,4-pentanedionato)titanium(IV) (ca. 63% in Isopropyl Alcohol), TCI America™
SDP

Catalog No. T2610500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2610500G 500 g
T261025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2610500G Supplier TCI America Supplier No. T2610500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17501-79-0
Molecular Formula C20H32O8Ti
Molecular Weight (g/mol) 448.335
MDL Number MFCD00045004
InChI Key RYSXWUYLAWPLES-MTOQALJVSA-N
Synonym Acetylacetone Titanium(IV) Salt, Titanium(IV) Acetylacetonate
PubChem CID 44629996
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;titanium
SMILES CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.[Ti]

Specifications

Quantity 500 g
UN Number 1219
Formula Weight 444.30
Physical Form Liquid
Chemical Name or Material Tetrakis(2,4-pentanedionato)titanium(IV) (ca. 63% in Isopropyl Alcohol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.