Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich TETRAKIS(4-BROMOPHENYL) SILANE ≥96%

Catalog No. 9140371G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-403-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-403-71G Supplier Sigma Aldrich Supplier No. 9140371G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrakis(bromophenyl) silane is an intermediate for emerging hole transport materials based on a silicon core. Materials containing this core have been shown to compete with SpiroMeOTAD for efficiency and stability in perovskite solar cells.

Specifications

CAS 18733-98-7
Molecular Formula C24H16Br4Si
Linear Formula C24H16Br4Si
Quantity 1 g
MDL Number MFCD30543388
Synonym 1,1 ,1 ,1 -Silanetetrayltetrakis[4-bromobenzene]; SiTPBr4; Tetra(4-bromophenyl)silane
Molecular Weight (g/mol) 652.09
Percent Purity ≥96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.