Learn More
Medchemexpress LLC Tetrakis (4-carboxyphenyl) porphyrin | 14609-54-2 | 98.29% | 790.79 | 100 MG
SDP
Catalog No. 5000366417
Encompass_Preferred
Tetrakis (4-carboxyphenyl) porphyrin (TCPP) is a chemical compound that functions as a metal remover. It is also known for its adsorption and photocatalytic performance, making it suitable for various research applications.
  • Acts as a metal remover
  • Exhibits adsorption performance on nano-TiO2 electrodes
  • Shows photocatalytic performance on nano-TiO2 electrodes
  • Suitable for solar energy conversion research
Catalog No. 50-003-66417 Supplier Medchemexpress LLC Supplier No. HYW008852100MG
Tetrakis (4-carboxyphenyl) porphyrin (TCPP) is a chemical compound that functions as a metal remover. It is also known for its adsorption and photocatalytic performance, making it suitable for various research applications.
  • Acts as a metal remover
  • Exhibits adsorption performance on nano-TiO2 electrodes
  • Shows photocatalytic performance on nano-TiO2 electrodes
  • Suitable for solar energy conversion research