Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrakis(Acetonitrile)Copper(I) Tetrafluoroborate

Catalog No. 6778925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-789-25G 5 g
67-789-21G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 67-789-25G Supplier Sigma Aldrich Supplier No. 6778925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.Catalyst in Diels-Alder and Ullmann reactions, intramolecular aromatic annulations, ring expansions in steroids, asymmetric aziridination of alkenes, substitution of allylic halides and mesylates, the Kharasch-Sosnovsky reaction.

Specifications

CAS 15418-29-8
Molecular Formula C8H12BCuF4N4
Linear Formula C8H12BCuF4N4
MDL Number MFCD09265110
Quantity 5 g
Synonym Cu(MeCN)4BF4; Copper(I) tetra(acetonitrile) tetrafluoroborate; Tetra(acetonitrile)copper(1+) tetrafluoroborate
Molecular Weight (g/mol) 314.56 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.