Learn More
Medchemexpress LLC Tetrakis(acetonitrile)palladium(II) tetrafluoroborate | 21797-13-7 | MFCD00043297 | >98.0% | 444.22 g/mol | C8H12B2F8N4Pd | 100 MG
SDP
Catalog No. 5000301553
Encompass_Preferred
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate is a palladium(II) complex provided as a solid reagent for laboratory research. It functions as a soluble Pd(II) source for homogeneous catalysis and organometallic synthesis, commonly used in cross-coupling reactions where a well-defined palladium species is required. The reagent is supplied with high purity and appropriate small-scale packaging for experimental work.
  • Soluble Pd(II) source in acetonitrile for homogeneous catalysis.
  • High purity suitable for research applications.
  • Stable solid reagent with light yellow to brown appearance.
  • Convenient small-scale packaging for laboratory use.
  • Useful in cross-coupling and organometallic synthesis.
Catalog No. 50-003-01553 Supplier Medchemexpress LLC Supplier No. HYW033277100MG
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate is a palladium(II) complex provided as a solid reagent for laboratory research. It functions as a soluble Pd(II) source for homogeneous catalysis and organometallic synthesis, commonly used in cross-coupling reactions where a well-defined palladium species is required. The reagent is supplied with high purity and appropriate small-scale packaging for experimental work.
  • Soluble Pd(II) source in acetonitrile for homogeneous catalysis.
  • High purity suitable for research applications.
  • Stable solid reagent with light yellow to brown appearance.
  • Convenient small-scale packaging for laboratory use.
  • Useful in cross-coupling and organometallic synthesis.