Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrakis(Decyl)Ammonium Bromide

Catalog No. 8758050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
87-580-50G 500 g
87-580-10G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 87-580-50G Supplier Sigma Aldrich Supplier No. 8758050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrakis(decyl)ammonium bromide is a cationic surfactant.Tetrakis(decyl)ammonium bromide acts as a phase transfer agent to transfer platinum salt from aqueous to organic phase during the synthesis of platinum nanocrystals. It plays the same role for transferring gold ions from an aqueous to an organic phase during the synthesis of gold nanoparticles.

Specifications

CAS 10424-65-4
Melting Point 67°C to 70°C
Molecular Formula C4H13NO -+ 5H2O
Linear Formula (CH3)4N(OH) -+ 5H2O
MDL Number MFCD00149566
Quantity 500 g
Synonym TDAB; Tetra-decylammonium bromide
Molecular Weight (g/mol) 181.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.