Learn More
Strem, An Ascensus Company CAS# 19824-55-6. 1g. Tetrakis(diethylamino)hafnium, 99% (99.99+%-Hf, <0.2% Zr) PURATREM. MFCD00799095
SDP
Supplier:  Strem, An Ascensus Company 7277501G
Encompass
CAS# 19824-55-6. 1g. Tetrakis(diethylamino)hafnium, 99% (99.99+%-Hf, <0.2% Zr) PURATREM. MFCD00799095. Molecular Weight: 467.01. Molecular Formula: Hf[N(CH2CH3)2]4. Color/form: light yellow liq. Strem# 72-7750. http://www.strem.com/catalog/v/72-7750/
Catalog No. 50-374-2