Learn More
Strem, An Ascensus Company CAS# 13801-49-5. 5g. Tetrakis(diethylamino)zirconium, 99%. MFCD00015649
SDP
Catalog No. 5090115281
Encompass
CAS# 13801-49-5. 5g. Tetrakis(diethylamino)zirconium, 99%. MFCD00015649. Molecular Weight: 379.74. Molecular Formula: Zr[N(CH2CH3)2]4. Color/form: yellow liq. Strem# 93-4040. http://www.strem.com/catalog/v/93-4040/
Catalog No. 50-901-15281 Supplier Strem, An Ascensus Company Supplier No. 9340405G
CAS# 13801-49-5. 5g. Tetrakis(diethylamino)zirconium, 99%. MFCD00015649. Molecular Weight: 379.74. Molecular Formula: Zr[N(CH2CH3)2]4. Color/form: yellow liq. Strem# 93-4040. http://www.strem.com/catalog/v/93-4040/