Learn More
Strem, An Ascensus Company TDMAZ 99% 25g
SDP
Catalog No. 5090111624
Encompass
CAS# 19756-04-8. 25g. Tetrakis(dimethylamino)zirconium(IV), 99% TDMAZ. MFCD00239502. Molecular Weight: 267.53. Molecular Formula: Zr[N(CH3)2]4. Color/form: white to light yellow xtl. Strem# 40-4100. http://www.strem.com/catalog/v/40-4100/
Catalog No. 50-901-11624 Supplier Strem, An Ascensus Company Supplier No. 40410025G
CAS# 19756-04-8. 25g. Tetrakis(dimethylamino)zirconium(IV), 99% TDMAZ. MFCD00239502. Molecular Weight: 267.53. Molecular Formula: Zr[N(CH3)2]4. Color/form: white to light yellow xtl. Strem# 40-4100. http://www.strem.com/catalog/v/40-4100/