Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(dimethylsilyl)silane 95.0+%, TCI America™
SDP

Catalog No. T17531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17531G Supplier TCI America Supplier No. T17531G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2003-85-2
Molecular Formula C8H28Si5
MDL Number MFCD01631315

Specifications

Color White
Quantity 1 g
UN Number 1325
Formula Weight 264.74
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Tetrakis(dimethylsilyl)silane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.