Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(hydroxymethyl)phosphonium Chloride (ca. 80% in Water), TCI America™
SDP

Catalog No. T060725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T060725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T060725ML Supplier TCI America Supplier No. T060725ML

missing translation for 'validMainframeMsg'

Specifications

CAS 124-64-1
Color Colorless
pH 3.5 - 4.5
Molecular Formula C4H12ClO4P
MDL Number MFCD00031687
UN Number 2922
Quantity 25 mL
Synonym tetrakis hydroxymethyl phosphonium chloride, thpc, pyroset tkc, retardol c, tetramethylolphosphonium chloride, tetrakis hydroxymethyl phosphanium chloride, unii-58wb2xcf8i, proban cc, tetrakis hydroxymethyl phosphochloride, ccris 317
InChI Key AKXUUJCMWZFYMV-UHFFFAOYSA-M
SMILES [Cl-].OC[P+](CO)(CO)CO
IUPAC Name tetrakis(hydroxymethyl)phosphanium chloride
Molecular Weight (g/mol) 190.56
PubChem CID 31298
Formula Weight 190.56
Physical Form Liquid
Chemical Name or Material Tetrakis(hydroxymethyl)phosphonium Chloride (ca. 80% in Water)
Show More Show Less
EINECSNumber (2)-2003
RTECSNumber TA2450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.