Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrakis(Hydroxymethyl)Phosphonium Chloride Solution

Catalog No. 404861500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-486-1500ML 500 mL
40-486-1100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 40-486-1500ML Supplier Sigma Aldrich Supplier No. 404861500ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrakis(hydroxymethyl)phosphonium chloride solution (80% in H2O) has been used as a reducing agent and stabilizing ligand for the synthesis of gold nanoparticles (AuNPs) fro mgold(III) chloride trihydrate (HAuCl4.3H2O). It is a tetra-functional, amine-reactive, aqueous crosslinker that can be used for tuning the properties of protein-based hydrogels for 3D cell encapsulation applications.

Specifications

CAS 124-64-1
Density 1.341 g/mL at 25°C
Molecular Formula C4H12ClO4P
Refractive Index n20/D 1.512
Linear Formula (HOCH2)4PCl
MDL Number MFCD00031687
Quantity 500 mL
Synonym THPC solution
Molecular Weight (g/mol) 190.56 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.