Learn More
Strem, An Ascensus Company CAS# 14221-02-4. 1g. Tetrakis(triphenylphosphine)platinum(0), 98%. MFCD00010014
SDP
Catalog No. 5090113339
Encompass
CAS# 14221-02-4. 1g. Tetrakis(triphenylphosphine)platinum(0), 98%. MFCD00010014. Molecular Weight: 1244.21. Molecular Formula: Pt[(C6H5)3P]4. Color/form: yellow xtl. Strem# 78-2030. http://www.strem.com/catalog/v/78-2030/
Catalog No. 50-901-13339 Supplier Strem, An Ascensus Company Supplier No. 7820301G
CAS# 14221-02-4. 1g. Tetrakis(triphenylphosphine)platinum(0), 98%. MFCD00010014. Molecular Weight: 1244.21. Molecular Formula: Pt[(C6H5)3P]4. Color/form: yellow xtl. Strem# 78-2030. http://www.strem.com/catalog/v/78-2030/