Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific Tetrakis(triphenylphosphine)palladium(0), 14221-01-3, MFCD00010012, 5g

Catalog No. 500119190
Encompass_Preferred

Molecular Formula C72H60P4Pd, Molecular Weight 1155.58, Melting Point 95-105 (dec)°

Catalog No. 50-011-9190 Supplier Matrix Scientific Supplier No. 0063955G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C72H60P4Pd, Molecular Weight 1155.58, Melting Point 95-105 (dec)°

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.