Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrakis(Triphenylphosphine)Palladium(0)

Catalog No. 21666625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
4 x 25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-666-625G 25 g
21-666-65G 5 g
21-666-64X25G 4 x 25 g
21-666-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Warning
Catalog No. 21-666-625G Supplier Sigma Aldrich Supplier No. 21666625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrakis(triphenylphosphine)palladium(0) can be used as a catalyst in:Negishi coupling (eq. 1), Suzuki coupling (eq. 2), Stille coupling (eq. 3), Sonogashira coupling reaction (eq. 4), and Buchwald-Hartwig amination reaction (eq. 5)The carbonylation of vinyl iodides (eq. 6)The reduction reaction of aryl bromides (eq. 7)Carbon-tin bond formation (eq. 8)

Specifications

CAS 14221-01-3
Molecular Formula C72H60P4Pd
Linear Formula Pd[(C6H5)3P]4
MDL Number MFCD00010012
Quantity 25 g
Synonym Palladium-tetrakis(triphenylphosphine); Pd(PPh3)4
Molecular Weight (g/mol) 1155.56 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.