Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetrakis(triphenylphosphine)platinum(0) | 14221-02-4 | MFCD00010014 | 97.0% | 1244.22 g·mol⁻1 | C72H60P4Pt | 250 MG
SDP

Catalog No. 5000303546
Encompass_Preferred

Tetrakis(triphenylphosphine)platinum(0) is an organometallic platinum(0) complex used as a reagent and precursor in organometallic synthesis and catalytic studies. It is typically handled as a light yellow to brown crystalline powder and is employed in research applications to prepare and study platinum-containing complexes.

  • High purity suitable for research applications (97.0%).
  • Molecular formula C72H60P4Pt and molecular weight 1244.22 g·mol⁻1.
  • Used as a precursor for platinum complexes and organometallic synthesis.
  • Commonly applied in catalytic studies and synthesis methodology development.
  • Typical appearance: light yellow to brown crystalline powder.

Catalog No. 50-003-03546 Supplier Medchemexpress LLC Supplier No. 250MGHYW034985
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrakis(triphenylphosphine)platinum(0) is an organometallic platinum(0) complex used as a reagent and precursor in organometallic synthesis and catalytic studies. It is typically handled as a light yellow to brown crystalline powder and is employed in research applications to prepare and study platinum-containing complexes.

  • High purity suitable for research applications (97.0%).
  • Molecular formula C72H60P4Pt and molecular weight 1244.22 g·mol⁻1.
  • Used as a precursor for platinum complexes and organometallic synthesis.
  • Commonly applied in catalytic studies and synthesis methodology development.
  • Typical appearance: light yellow to brown crystalline powder.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.