Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetrakis(triphenylphosphine)platinum(0) | 14221-02-4 | MFCD00010014 | 98.2% | 1244.22 g/mol | C72H60P4Pt | 100 MG
SDP

Catalog No. 5000300175
Encompass_Preferred

Tetrakis(triphenylphosphine)platinum(0) is an organometallic platinum(0) complex used as a reagent and catalyst in organic and organometallic synthesis. Supplied as a light yellow solid with high purity, it is intended for research applications in academic and industrial laboratories.

  • High purity (98.2%) suitable for research use.
  • Organometallic platinum(0) catalyst precursor for homogeneous reactions.
  • Molecular formula C72H60P4Pt; mol wt 1244.22 g/mol.
  • Appearance: light yellow solid; protect from light and store under inert atmosphere.
  • Supplied with certificate of analysis and safety data sheet for quality and handling information.

Catalog No. 50-003-00175 Supplier Medchemexpress LLC Supplier No. HYW034985100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrakis(triphenylphosphine)platinum(0) is an organometallic platinum(0) complex used as a reagent and catalyst in organic and organometallic synthesis. Supplied as a light yellow solid with high purity, it is intended for research applications in academic and industrial laboratories.

  • High purity (98.2%) suitable for research use.
  • Organometallic platinum(0) catalyst precursor for homogeneous reactions.
  • Molecular formula C72H60P4Pt; mol wt 1244.22 g/mol.
  • Appearance: light yellow solid; protect from light and store under inert atmosphere.
  • Supplied with certificate of analysis and safety data sheet for quality and handling information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.