Learn More
Medchemexpress LLC Tetrakis(triphenylphosphine)platinum(0) | 14221-02-4 | MFCD00010014 | 98.2% | 1244.22 g·mol⁻¹ | C72H60P4Pt | 500 MG
SDP
Catalog No. 5000301545
Encompass_Preferred
Tetrakis(triphenylphosphine)platinum is a platinum(0) coordination complex used as a reagent and precursor to platinum complexes in organometallic and organic synthesis. Supplied as a light yellow solid, it is typically handled under inert atmosphere and protected from light for storage and use in research.
  • Coordination complex used as a platinum(0) reagent.
  • Light yellow solid; handle under inert gas and protect from light.
  • High purity suitable for research and synthetic applications.
  • Molecular formula C72H60P4Pt; molecular weight 1244.22 g·mol⁻¹.
  • Available in small laboratory pack sizes for bench-scale work.
Catalog No. 50-003-01545 Supplier Medchemexpress LLC Supplier No. HYW034985500MG
Tetrakis(triphenylphosphine)platinum is a platinum(0) coordination complex used as a reagent and precursor to platinum complexes in organometallic and organic synthesis. Supplied as a light yellow solid, it is typically handled under inert atmosphere and protected from light for storage and use in research.
  • Coordination complex used as a platinum(0) reagent.
  • Light yellow solid; handle under inert gas and protect from light.
  • High purity suitable for research and synthetic applications.
  • Molecular formula C72H60P4Pt; molecular weight 1244.22 g·mol⁻¹.
  • Available in small laboratory pack sizes for bench-scale work.