Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrakis(Triphenylphosphine)Platinum(0)

Catalog No. 2449615G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-496-15G 5 g
24-496-11G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-496-15G Supplier Sigma Aldrich Supplier No. 2449615G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrakis(triphenylphosphine)platinum(0) is used as a precursor to platinum complexes.It can undergo oxidative addition to organochalcogen compounds such as dithienyl ditelluride through the cleavage of the carbon-chalcogen bond. This reaction can play a role in homogeneous catalysis.

Specifications

CAS 14221-02-4
Molecular Formula C72H60P4Pt
Linear Formula Pt[(C6H5)3P]4
Quantity 5 g
MDL Number MFCD00010014
Synonym Pt(PPh3)4; Platinum(0)-tetrakis(triphenylphosphine)
Molecular Weight (g/mol) 1244.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.