Learn More
Medchemexpress LLC Tetrakis(triphenylphosphine)platinum | 14221-02-4 | MFCD00010014 | 98.2% | 1244.22 g·mol⁻¹ | C72H60P4Pt | 1 G
SDP
Catalog No. 5000299845
Encompass_Preferred
Tetrakis(triphenylphosphine)platinum is a platinum(0) organometallic complex used as a reagent and catalyst precursor in organic and organometallic synthesis. It is supplied as a light yellow to yellow solid for research use, with high specified purity and defined molecular composition, suitable for laboratory-scale catalytic and synthetic applications.
  • Platinum(0) complex useful as a catalyst precursor.
  • High purity for reproducible experimental results.
  • Stable, isolable solid convenient for handling and storage.
  • Applicable to hydrosilylation, oxidative addition, and related transformations.
  • Available in small lab-scale packaging for research use.
Catalog No. 50-002-99845 Supplier Medchemexpress LLC Supplier No. HYW0349851G
Tetrakis(triphenylphosphine)platinum is a platinum(0) organometallic complex used as a reagent and catalyst precursor in organic and organometallic synthesis. It is supplied as a light yellow to yellow solid for research use, with high specified purity and defined molecular composition, suitable for laboratory-scale catalytic and synthetic applications.
  • Platinum(0) complex useful as a catalyst precursor.
  • High purity for reproducible experimental results.
  • Stable, isolable solid convenient for handling and storage.
  • Applicable to hydrosilylation, oxidative addition, and related transformations.
  • Available in small lab-scale packaging for research use.