Learn More
AdipoGen Tetrakistrimethylsilylsilane
SDP
Catalog No. 502559937
Encompass_Preferred
Chemical. CAS 4098-98-0. Formula CH33Si4Si. MW BD9837. Tetrakistrimethylsilylsilane TTMSS is an organosilicon compound. TTMSS is widely used as a reagent and precursor in organic synthesis and silicon-based chemistry. TTMSS is commonly used as a protecting group for various functional groups in organic synthesis, in deoxygenation reactions to remove oxygen-containing functional groups like ketones, aldehydes, and esters, converting them into the corresponding trimethylsilyl ethers. TTMSS is utilized in silylation reactions to introduce trimethylsilyl groups onto organic compounds, enhancing their volatility and stability for gas chromatography and mass spectrometry analysis. Due to its diverse applications in organic chemistry, Tetrakistrimethylsilylsilane has become a valuable tool for synthetic chemists, particularly in the fields of silicon-based chemistry, protecting group strategies, and cross-coupling reactions.
Catalog No. 50-255-9937 Supplier AdipoGen Supplier No. CDXT0492G005
Chemical. CAS 4098-98-0. Formula CH33Si4Si. MW BD9837. Tetrakistrimethylsilylsilane TTMSS is an organosilicon compound. TTMSS is widely used as a reagent and precursor in organic synthesis and silicon-based chemistry. TTMSS is commonly used as a protecting group for various functional groups in organic synthesis, in deoxygenation reactions to remove oxygen-containing functional groups like ketones, aldehydes, and esters, converting them into the corresponding trimethylsilyl ethers. TTMSS is utilized in silylation reactions to introduce trimethylsilyl groups onto organic compounds, enhancing their volatility and stability for gas chromatography and mass spectrometry analysis. Due to its diverse applications in organic chemistry, Tetrakistrimethylsilylsilane has become a valuable tool for synthetic chemists, particularly in the fields of silicon-based chemistry, protecting group strategies, and cross-coupling reactions.