Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetralin™ 1,2,3,4-Tetrahydronaphthalene, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019997 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9997 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9997 Supplier MilliporeSigma Supelco Supplier No. 954625MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line Tetralin
Ignition Point 723°F
CAS 119-64-2
Vapor Pressure 0.18 mmHg (20°C)
Vapor Density 4.55 (vs. air)
Density 0.973 g/mL (at 25°C)
Flash Point 71°C
Physical Form Neat
Linear Formula C10H12
MDL Number MFCD00001733
Melting Point -35°C
Boiling Point 207°C
Synonym Tetralin(R) solvent
Type Analytical Standard
Quantity 5 mL
Formula Weight 132.20
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less

Tetralin is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.