Learn More
AdipoGen TetraMe-1 8-naphthalindiamine
SDP
Catalog No. 502559928
Encompass_Preferred
Chemical. CAS 20734-58-1. Formula C14H18N2. MW 214.31. 1,8-Bisdimethylaminonaphthalene Proton Sponge is an organic compound that has several very interesting properties. One is its very high basicity, another is its spectroscopic properties. It is a very strong base with weak nucleophilic character due to steric effects. Due to this it has been used in organic synthesis as a highly selective non-nucleophilic base. Proton sponge also exhibits a very high affinity for boron, and is capable of displacing hydride from borane to form a boronium-borohydride ion pair.
Catalog No. 50-255-9928 Supplier AdipoGen Supplier No. CDXT0909G010
Chemical. CAS 20734-58-1. Formula C14H18N2. MW 214.31. 1,8-Bisdimethylaminonaphthalene Proton Sponge is an organic compound that has several very interesting properties. One is its very high basicity, another is its spectroscopic properties. It is a very strong base with weak nucleophilic character due to steric effects. Due to this it has been used in organic synthesis as a highly selective non-nucleophilic base. Proton sponge also exhibits a very high affinity for boron, and is capable of displacing hydride from borane to form a boronium-borohydride ion pair.