Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetramethylammonium Hydrogen Sulfate Monohydrate

Catalog No. 8772650G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
87-726-50G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 87-726-50G Supplier Sigma Aldrich Supplier No. 8772650G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Crystals of tetramethylammonium hydrogen sulfate monohydrate generally exhibit ferroelectricity in the temperature range from -104 C to 40 C.Tetramethylammonium hydrogen sulfate monohydrate can be used as a phase transfer catalyst in polymer synthesis.

Specifications

CAS 75-76-3
Melting Point -99°C
Density 0.648 g/mL at 25°C
Boiling Point 26°C to 28°C
Molecular Formula C4H12Si
Linear Formula Si(CH3)4
Quantity 50 g
MDL Number MFCD00008274
Molecular Weight (g/mol) 88.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.