Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethyl 1,2,3,4-Cyclobutanetetracarboxylate 98.0+%, TCI America™
SDP

Catalog No. T27695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27695G Supplier TCI America Supplier No. T27695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crystal to powder reagent for laboratory research applications.

Chemical Identifiers

CAS 14495-41-1
Molecular Formula C12H16O8
Molecular Weight (g/mol) 288.25
MDL Number MFCD00013269
InChI Key NXMOMNBIDWYNOP-UHFFFAOYSA-N
Synonym 1,2,3,4-Cyclobutanetetracarboxylic Acid Tetramethyl Ester
PubChem CID 266035
IUPAC Name 1,2,3,4-tetramethyl cyclobutane-1,2,3,4-tetracarboxylate
SMILES COC(=O)C1C(C(C1C(=O)OC)C(=O)OC)C(=O)OC

Specifications

Melting Point 147°C
Color White
Quantity 5 g
Formula Weight 288.25
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Tetramethyl 1,2,3,4-Cyclobutanetetracarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.