Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethyl-1,4-benzoquinone 98.0+%, TCI America™
SDP

Catalog No. T06721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06721G 1 g
T06725G 5 g
T067225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T06721G Supplier TCI America Supplier No. T06721G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 527-17-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00001604
InChI Key WAMKWBHYPYBEJY-UHFFFAOYSA-N
Synonym duroquinone, tetramethyl-1,4-benzoquinone, tetramethyl-p-benzoquinone, tetramethylquinone, 2,3,5,6-tetramethyl-1,4-benzoquinone, tetramethyl-p-quinone, 2,3,5,6-tetramethylbenzoquinone, p-benzoquinone, tetramethyl, p-benzoquinone, 2,3,5,6-tetramethyl, 2,3,5,6-tetramethyl-p-benzoquinone
PubChem CID 68238
ChEBI CHEBI:42023
IUPAC Name 2,3,5,6-tetramethylcyclohexa-2,5-diene-1,4-dione
SMILES CC1=C(C(=O)C(=C(C1=O)C)C)C

Specifications

Melting Point 112°C
Color Yellow
Quantity 1 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethyl-1,4-benzoquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.