Learn More
Medchemexpress LLC Tetramethylammonium 10g
SDP
Catalog No. 5000206903
Encompass_Preferred
Tetramethylammonium p-toluenesulfonate is a quaternary ammonium compound belonging to the class of alkylammonium salts This compound is widely used as a phase transfer catalyst in organic synthesis to facilitate the transfer of reactants between immiscible phases Furthermore it can be used as a surfactant emulsifier or stabilizer in various industrial applications Its unique chemical properties make it an important ingredient in the production of pharmaceuticals agrochemicals and materials science products
Catalog No. 50-002-06903 Supplier Medchemexpress LLC Supplier No. HYW10040410G
Tetramethylammonium p-toluenesulfonate is a quaternary ammonium compound belonging to the class of alkylammonium salts This compound is widely used as a phase transfer catalyst in organic synthesis to facilitate the transfer of reactants between immiscible phases Furthermore it can be used as a surfactant emulsifier or stabilizer in various industrial applications Its unique chemical properties make it an important ingredient in the production of pharmaceuticals agrochemicals and materials science products