Learn More
Medchemexpress LLC Tetramethylammonium 5g
SDP
Catalog No. 5000206202
Encompass_Preferred
Tetramethylammonium p-toluenesulfonate is a quaternary ammonium compound belonging to the class of alkylammonium salts This compound is widely used as a phase transfer catalyst in organic synthesis to facilitate the transfer of reactants between immiscible phases Furthermore it can be used as a surfactant emulsifier or stabilizer in various industrial applications Its unique chemical properties make it an important ingredient in the production of pharmaceuticals agrochemicals and materials science products
Catalog No. 50-002-06202 Supplier Medchemexpress LLC Supplier No. HYW1004045G
Tetramethylammonium p-toluenesulfonate is a quaternary ammonium compound belonging to the class of alkylammonium salts This compound is widely used as a phase transfer catalyst in organic synthesis to facilitate the transfer of reactants between immiscible phases Furthermore it can be used as a surfactant emulsifier or stabilizer in various industrial applications Its unique chemical properties make it an important ingredient in the production of pharmaceuticals agrochemicals and materials science products