Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetramethylammonium acetate hydrate | 1266615-86-4 | MFCD03066126 | 97.0% | 151.20 g/mol | C6H17NO3 | 500 G
SDP

Catalog No. 5000283119
Encompass_Preferred

Tetramethylammonium acetate hydrate is supplied as a biochemical reagent for life-science research and is intended for research use only. The compound is identified by CAS 1266615-86-4, with molecular formula C6H17NO3 and molecular weight 151.20 g/mol.

  • Biochemical reagent suitable for life-science research
  • CAS number 1266615-86-4
  • Molecular formula C6H17NO3
  • Molecular weight 151.20 g/mol
  • Available as a 500 G package (verify with manufacturer)
  • Provided for research use only, not for clinical use

Catalog No. 50-002-83119 Supplier Medchemexpress LLC Supplier No. HYW130431500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetramethylammonium acetate hydrate is supplied as a biochemical reagent for life-science research and is intended for research use only. The compound is identified by CAS 1266615-86-4, with molecular formula C6H17NO3 and molecular weight 151.20 g/mol.

  • Biochemical reagent suitable for life-science research
  • CAS number 1266615-86-4
  • Molecular formula C6H17NO3
  • Molecular weight 151.20 g/mol
  • Available as a 500 G package (verify with manufacturer)
  • Provided for research use only, not for clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.