Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium borohydride, 95%, Thermo Scientific™

Catalog No. AA3257404
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3257404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3257404 Supplier Thermo Scientific Chemicals Supplier No. 3257404

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16883-45-7
Molecular Formula C4H12BN
Molecular Weight (g/mol) 84.96
MDL Number MFCD00011778
InChI Key TVQBZLPZXCVNEO-UHFFFAOYSA-N
Synonym tetramethylammonium borohydride, borohydride, polymer-supported, tetramethylammonium borohydrate, tetramethylammonium tetrahydroborate 1-, tetramethylammonium tetrahydroborate, tetra-methyl ammonium boron hydride, boron 1-; tetramethylazanium, n,n,n-trimethylmethanaminium tetrahydroborate 1-, ammonium, tetramethyl-, tetrahydroborate 1-, methanaminium, n,n,n-trimethyl-, tetrahydroborate 1-
PubChem CID 6328186
IUPAC Name tetramethylazanium λ¹-boranuidylidene
SMILES [B-].C[N+](C)(C)C

Specifications

Melting Point 150°C (decomposition)(vac.)
Density 0.813g/mL
Quantity 2 g
UN Number UN1409
Formula Weight 88.99
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Tetramethylammonium borohydride
Hazard Category Danger
Hazard Statement GHS H Statement
H260-H300-H311-H315-H319-H335
In contact with water releases flammable gases which may ignite spontaneously.
Fatal if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P231+P232-P301+P310-P305+P351+P338-P361-P405-P501a
Handle under inert gas.
Protect from moisture.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: METAL HYDRIDES, WATER REACTIVE, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.