Learn More
Medchemexpress LLC Tetramethylammonium bromide | 64-20-0 | MFCD00011626 | 98.0% | 154.05 g/mol | C4H12BrN | 100g
SDP
Catalog No. 5000269202
Encompass_Preferred
Tetramethylammonium bromide is a quaternary ammonium salt commonly used as a surfactant phase transfer catalyst and template molecule in various chemical reactions especially in organic synthesis and materials science Tetramethylammonium bromide has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes
Catalog No. 50-002-69202 Supplier Medchemexpress LLC Supplier No. HYY0658100G
Tetramethylammonium bromide is a quaternary ammonium salt commonly used as a surfactant phase transfer catalyst and template molecule in various chemical reactions especially in organic synthesis and materials science Tetramethylammonium bromide has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes