Learn More
Tetramethylammonium chloride, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 420.0°C |
| Color | White to Ivory |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% min. (Argentometry) |
| Packaging | Plastic bottle |
| Linear Formula | (CH3)4NCl |
| Quantity | 1 kg |
| Solubility Information | Solubility in water: >60% (20°C). Other solubilities: insoluble in ether, benzene, slightly soluble in ethanol |
| Vapor Pressure | Negligible |
| Formula Weight | 109.6 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Fatal if swallowed. Toxic in contact with skin. Causes skin irritation. Causes damage to organs if swallowed. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement IF exposed: Call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water |
| EINECSNumber | 200-880-8 |
| RTECSNumber | BS7700000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.