Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium Chloride, High Purity, 99%, Spectrum™ Chemical
SDP

Catalog No. 18614373 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-373 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-614-373 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T103650KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-57-0
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.60
InChI Key OKIZCWYLBDKLSU-UHFFFAOYSA-M
IUPAC Name tetramethylazanium chloride
SMILES [Cl-].C[N+](C)(C)C

Specifications

CAS Max % 1
Packaging Fiber Drum
Quantity 50 kg
Percent Purity 0.99
Grade High Purity

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.