Learn More
Medchemexpress LLC Tetramethylammonium-d12 (chloride) | 23789-03-9 | 99.9% | 121.67 | 100 MG
SDP
Catalog No. 5000451450
Encompass_Preferred
Tetramethylammonium-d12 (chloride) is the deuterium labeled Tetramethylammonium chloride. It is intended for research use only. This compound can be utilized as a tracer and as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS. Deuteration is notable for its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-51450 Supplier Medchemexpress LLC Supplier No. HYW012954S100MG
Tetramethylammonium-d12 (chloride) is the deuterium labeled Tetramethylammonium chloride. It is intended for research use only. This compound can be utilized as a tracer and as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS. Deuteration is notable for its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.