Learn More
Medchemexpress LLC Tetramethylammonium hexafluorophosphate | 558-32-7 | MFCD00012012 | 98.0% | 219.11 g/mol | C4H12F6NP | 25 G
SDP
Catalog No. 5000283519
Encompass_Preferred
Tetramethylammonium hexafluorophosphate is an inorganic salt used as a reagent and supporting electrolyte in electrochemical, synthetic, and biochemical research. It supplies the tetramethylammonium cation paired with the hexafluorophosphate anion and is provided as a solid for laboratory use.
  • Supplies tetramethylammonium cation and hexafluorophosphate anion.
  • Suitable as a supporting electrolyte for electrochemical applications.
  • Used in synthetic and biochemical research as a reagent.
  • Molecular weight 219.11 g/mol.
  • Available in laboratory pack sizes, including 25 g.
Catalog No. 50-002-83519 Supplier Medchemexpress LLC Supplier No. HYW12767325G
Tetramethylammonium hexafluorophosphate is an inorganic salt used as a reagent and supporting electrolyte in electrochemical, synthetic, and biochemical research. It supplies the tetramethylammonium cation paired with the hexafluorophosphate anion and is provided as a solid for laboratory use.
  • Supplies tetramethylammonium cation and hexafluorophosphate anion.
  • Suitable as a supporting electrolyte for electrochemical applications.
  • Used in synthetic and biochemical research as a reagent.
  • Molecular weight 219.11 g/mol.
  • Available in laboratory pack sizes, including 25 g.