Learn More
Medchemexpress LLC Tetramethylammonium hexafluorophosphate | 558-32-7 | MFCD00012012 | 99.0% | 219.11 g·mol⁻1 | C4H12F6NP | 50 G
SDP
Catalog No. 5000282591
Encompass_Preferred
Tetramethylammonium hexafluorophosphate is a tetraalkylammonium salt supplied as a white to off-white solid reagent used in organic synthesis and electrochemical applications. It serves as a stable source of tetramethylammonium cation and hexafluorophosphate anion for use as a supporting electrolyte, phase-transfer agent, or reagent in analytical workflows.
  • Supplied as a white to off-white solid.
  • Suitable for organic synthesis and electrochemical studies.
  • Stable and low hygroscopicity for reliable handling.
  • Available in laboratory pack sizes, including 50 g.
  • Provides tetramethylammonium and hexafluorophosphate ions for analytical use.
Catalog No. 50-002-82591 Supplier Medchemexpress LLC Supplier No. HYW12767350G
Tetramethylammonium hexafluorophosphate is a tetraalkylammonium salt supplied as a white to off-white solid reagent used in organic synthesis and electrochemical applications. It serves as a stable source of tetramethylammonium cation and hexafluorophosphate anion for use as a supporting electrolyte, phase-transfer agent, or reagent in analytical workflows.
  • Supplied as a white to off-white solid.
  • Suitable for organic synthesis and electrochemical studies.
  • Stable and low hygroscopicity for reliable handling.
  • Available in laboratory pack sizes, including 50 g.
  • Provides tetramethylammonium and hexafluorophosphate ions for analytical use.