Learn More
Medchemexpress LLC Tetramethylammonium hydrogensulfate | 80526-82-5 | MFCD00036149 | 171.22 g/mol | C4H13NO4S | 100 G
SDP
Catalog No. 5000209236
Encompass_Preferred
Tetramethylammonium hydrogensulfate is an organic salt used as a reagent in laboratory research that provides the tetramethylammonium cation paired with a hydrogensulfate (bisulfate) anion. It is used in synthesis and analytical workflows where tetramethylammonium salts are required.
  • Used as a reagent for organic synthesis and phase-transfer catalysis.
  • Suitable for ion-pair chromatography and other analytical applications.
  • Molecular formula C4H13NO4S and molecular weight 171.22 g/mol.
  • Available as a 100 G packaged reagent.
Catalog No. 50-002-09236 Supplier Medchemexpress LLC Supplier No. HYW040184100G
Tetramethylammonium hydrogensulfate is an organic salt used as a reagent in laboratory research that provides the tetramethylammonium cation paired with a hydrogensulfate (bisulfate) anion. It is used in synthesis and analytical workflows where tetramethylammonium salts are required.
  • Used as a reagent for organic synthesis and phase-transfer catalysis.
  • Suitable for ion-pair chromatography and other analytical applications.
  • Molecular formula C4H13NO4S and molecular weight 171.22 g/mol.
  • Available as a 100 G packaged reagent.