Learn More
Medchemexpress LLC Tetramethylammonium hydrogensulfate | 80526-82-5 | MFCD00036149 | 171.22 g/mol | C4H13NO4S | 50 G
SDP
Catalog No. 5000208904
Encompass_Preferred
Tetramethylammonium hydrogensulfate is an organic tetramethylammonium salt supplied as a laboratory reagent for chemical and life-science research applications. It provides a tetramethylammonium counterion and is commonly used in ion-pairing and analytical procedures.
  • Reagent for laboratory research and analytical methods.
  • Provides tetramethylammonium counterion useful in ion-pair chromatography.
  • Chemical formula C4H13NO4S, CAS 80526-82-5.
  • Sold as a 50 G unit for laboratory use.
Catalog No. 50-002-08904 Supplier Medchemexpress LLC Supplier No. HYW04018450G
Tetramethylammonium hydrogensulfate is an organic tetramethylammonium salt supplied as a laboratory reagent for chemical and life-science research applications. It provides a tetramethylammonium counterion and is commonly used in ion-pairing and analytical procedures.
  • Reagent for laboratory research and analytical methods.
  • Provides tetramethylammonium counterion useful in ion-pair chromatography.
  • Chemical formula C4H13NO4S, CAS 80526-82-5.
  • Sold as a 50 G unit for laboratory use.