Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetramethylammonium hydrogensulfate | 80526-82-5 | MFCD00036149 | >99.0% | 171.22 g/mol | C4H13NO4S | 10 G
SDP

Catalog No. 5000208661
Encompass_Preferred

Tetramethylammonium hydrogensulfate is a white to off-white solid inorganic salt used as an ion-pairing reagent and general-purpose reagent in analytical and biochemical applications. It has molecular formula C4H13NO4S and a molecular weight of 171.22 g/mol. Available in small-pack sizes for laboratory use.

  • White to off-white solid.
  • Molecular formula C4H13NO4S.
  • Molecular weight 171.22 g/mol.
  • CAS number 80526-82-5.
  • Available in 10 g, 25 g, 50 g, 100 g, and bulk quantities.
  • Product data sheet and SDS are available for handling and safety information.

Catalog No. 50-002-08661 Supplier Medchemexpress LLC Supplier No. HYW04018410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetramethylammonium hydrogensulfate is a white to off-white solid inorganic salt used as an ion-pairing reagent and general-purpose reagent in analytical and biochemical applications. It has molecular formula C4H13NO4S and a molecular weight of 171.22 g/mol. Available in small-pack sizes for laboratory use.

  • White to off-white solid.
  • Molecular formula C4H13NO4S.
  • Molecular weight 171.22 g/mol.
  • CAS number 80526-82-5.
  • Available in 10 g, 25 g, 50 g, 100 g, and bulk quantities.
  • Product data sheet and SDS are available for handling and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.