Learn More
Medchemexpress LLC Tetramethylammonium hydrogensulfate | 80526-82-5 | MFCD00150356 | ≥98.0% | 171.22 g/mol | C4H13NO4S | 25 G
SDP
Catalog No. 5000209143
Encompass_Preferred
Tetramethylammonium hydrogensulfate is a quaternary ammonium salt supplied as a white crystalline powder used as a source of tetramethylammonium in chemical syntheses and biochemical assays. It has formula C4H13NO4S and a molecular weight of 171.22 g/mol. The material is provided in laboratory-scale pack sizes with accompanying documentation for safe handling.
  • White crystalline powder, soluble in water.
  • Provides tetramethylammonium counterion for synthetic applications.
  • Suitable for phase-transfer catalysis and reagent preparation.
  • Supplied in laboratory-scale pack sizes for bench experiments.
  • Data sheet and safety data sheet available for handling guidance.
Catalog No. 50-002-09143 Supplier Medchemexpress LLC Supplier No. 25GHYW040184
Tetramethylammonium hydrogensulfate is a quaternary ammonium salt supplied as a white crystalline powder used as a source of tetramethylammonium in chemical syntheses and biochemical assays. It has formula C4H13NO4S and a molecular weight of 171.22 g/mol. The material is provided in laboratory-scale pack sizes with accompanying documentation for safe handling.
  • White crystalline powder, soluble in water.
  • Provides tetramethylammonium counterion for synthetic applications.
  • Suitable for phase-transfer catalysis and reagent preparation.
  • Supplied in laboratory-scale pack sizes for bench experiments.
  • Data sheet and safety data sheet available for handling guidance.