Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium Hydroxide, 10% Solution in Water, (U.S.P. Test Solution), Spectrum™ Chemical
SDP

Catalog No. 18607338 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-338 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-338 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T2449500MLGL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

T2449, 75-59-2

Spectrum™ Chemical Tetramethylammonium Hydroxide, 10 Percent Solution in Water, (U.S.P. Test Solution)

Chemical Identifiers

CAS 75-59-2 , 7732-18-5
Molecular Formula C4H13NO
Molecular Weight (g/mol) 91.15
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
IUPAC Name tetramethylazanium hydroxide
SMILES [OH-].C[N+](C)(C)C

Specifications

Color Colorless to Pale Yellow
CAS Max % 10%, 90%
Concentration 10%
Percent Purity 9 to 11%
Grade Reagent
Packaging Amber Glass Bottle
Quantity 500 mL
Chemical Name or Material Tetramethylammonium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.