Learn More
Medchemexpress LLC Tetramethylammonium iodide | 75-58-1 | MFCD00011629 | 98.0% | 201.05 g/mol | C4H12IN | 100 G
SDP
Catalog No. 5000302119
Encompass_Preferred
Tetramethylammonium iodide (CAS 75-58-1) is a quaternary ammonium salt supplied as a white to off-white solid (C4H12IN, MW 201.05 g/mol) used in research settings as a surfactant and reagent. It is intended for laboratory use and is typically provided with batch documentation for quality control.
  • High purity: 98.0%.
  • White to off-white solid form for ease of handling.
  • Reduces surface tension in aqueous solutions.
  • Suitable for biochemical assays and general laboratory reagent use.
  • Defined molecular weight: 201.05 g/mol.
  • Supplied with batch COA and datasheet for traceability.
Catalog No. 50-003-02119 Supplier Medchemexpress LLC Supplier No. HYW014861100G
Tetramethylammonium iodide (CAS 75-58-1) is a quaternary ammonium salt supplied as a white to off-white solid (C4H12IN, MW 201.05 g/mol) used in research settings as a surfactant and reagent. It is intended for laboratory use and is typically provided with batch documentation for quality control.
  • High purity: 98.0%.
  • White to off-white solid form for ease of handling.
  • Reduces surface tension in aqueous solutions.
  • Suitable for biochemical assays and general laboratory reagent use.
  • Defined molecular weight: 201.05 g/mol.
  • Supplied with batch COA and datasheet for traceability.