Learn More
Medchemexpress LLC Tetramethylammonium tetrafluoroborate | 661-36-9 | MFCD00011745 | 98.0% | 160.95 | C4H12BF4N | 25 G
SDP
Catalog No. 5000282668
Encompass_Preferred
Tetramethylammonium tetrafluoroborate is a quaternary ammonium salt supplied as a white to off-white solid and used as an electrolyte, phase-transfer reagent, and general-purpose reagent in organic and electrochemical research applications.
  • High chemical purity suitable for research use.
  • Stable tetrafluoroborate counterion for electrochemical applications.
  • Available in multiple pack sizes to support different scales of work.
  • Solid form that is easy to weigh, handle, and store under standard laboratory conditions.
  • Well-known identifiers: molecular weight 160.95, formula C4H12BF4N, CAS 661-36-9.
Catalog No. 50-002-82668 Supplier Medchemexpress LLC Supplier No. HYW12767225G
Tetramethylammonium tetrafluoroborate is a quaternary ammonium salt supplied as a white to off-white solid and used as an electrolyte, phase-transfer reagent, and general-purpose reagent in organic and electrochemical research applications.
  • High chemical purity suitable for research use.
  • Stable tetrafluoroborate counterion for electrochemical applications.
  • Available in multiple pack sizes to support different scales of work.
  • Solid form that is easy to weigh, handle, and store under standard laboratory conditions.
  • Well-known identifiers: molecular weight 160.95, formula C4H12BF4N, CAS 661-36-9.